Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LAKCLGPFDEW ESRLRVAKES GYNMIHFTPL QTLGLSRSCY SLANQLELNP DFSRPNRKYT WNDVGQLVEK LKKEWNVICI TDVVYNHTAA NSKWIQEHPE
Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Apr; 4-alpha-glucanotransferase; 4-glucantransferase; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; Amylo-1,6-glucosidase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Amylo-alpha-1,6-glucosidase; Dextrin 6-alpha-D-glucosidase; Glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1; Oligo-1,4-1,4-glucantransferase
Gene Aliases: AGL; GDE
UniProt ID: (Human) P35573
Entrez Gene ID: (Human) 178
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support