Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSFSASLGRAG GVSAQRDLFE ERGEEYLSAF DKKAQADFDS CISSQRIGQE LLFPPQENVQ DAGAPGGHTQ NLRCSPLEPD FVPDEKKPEC GSWDVSHQP
Highest antigen sequence identity to the following orthologs - mouse 86%, rat 87%.
PRRC2B (Proline Rich Coiled-Coil 2B) is a Protein Coding gene. An important paralog of this gene isPRRC2C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLA-B associated transcript 2-like 1; HLA-B-associated transcript 2-like 1; proline-rich coiled-coil 2B; Proline-rich coiled-coil protein 2B; protein BAT2-like 1; Protein PRRC2B
Gene Aliases: BAT2L; BAT2L1; KIAA0515; LQFBS-1; PRRC2B
UniProt ID: (Human) Q5JSZ5
Entrez Gene ID: (Human) 84726
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support