Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQVVPFSQLA DAAADFTLLA MDMPSTAPSD FQPQPVISII QLFGWDDIIC PQVVARYLSH VLQNSTLCEA LSHSGYVSFQ ALTVRSWIRC
Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.
Component of the MMS22L-TONSL complex, a complex that stimulates the recombination-dependent repair of stalled or collapsed replication forks. The MMS22L-TONSL complex is required to maintain genome integrity during DNA replication by promoting homologous recombination-mediated repair of replication fork-associated double-strand breaks. It may act by mediating the assembly of RAD51 filaments on ssDNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Methyl methanesulfonate-sensitivity protein 22-like; MMS22-like, DNA repair protein; Protein MMS22-like
Gene Aliases: C6orf167; dJ39B17.2; MMS22L
UniProt ID: (Human) Q6ZRQ5
Entrez Gene ID: (Human) 253714
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support