Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: 293T, HeLa, NIH/3T3, HL-60, BT-474, HT-29; Cellular Location: Cytoplasm
Immunogen sequence: ICRPSGRKSS KMQAFRIWDV NQKTFYLRNN QLVAGYLQGP NVNLEEKIDV VPIEPHALFL GIHGGKMCLS CVKSGDETRL QLEAVNITDL SENRKQDKRF AFIRSDSGPT TSFESAACPG WFLCTAMEAD QPVSLTNMPD EGVMVTKFYF QEDE
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anakinra; CD121A; D2S1473; ICIL-1RA; IL 1 receptor antagonist; il-1 receptor; il-1r; IL-1RN; IL1 inhibitor; il1 receptor; IL1 receptor antagonist; IL1-RA; IL1R; IL1RN (IL1F3); interleukin-1 receptor; Interleukin-1 receptor antagonist protein; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); IRAP; MGC10430; P80; type II interleukin-1 receptor antagonist
Gene Aliases: DIRA; F630041P17Rik; ICIL-1RA; IL-1ra; IL-1ra3; IL-1RN; IL1F3; IL1RA; IL1RN; IRAP; MVCD4
UniProt ID: (Human) P18510, (Mouse) P25085, (Rat) P25086
Entrez Gene ID: (Human) 3557, (Mouse) 16181, (Rat) 60582
Molecular Function: interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support