Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PEKQESHQKS HGPGSDLENF ALFKLEEREK KVYPKAVERS STFENKASMQ EAPPLGSLLK DALHKQASVR ASEGAMSDGP VPAEHRQGGG DFRRAPAPGT LQDGLCHS
Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine(Ser/Thr) protein kinases. MAST4 (microtubule associated serine/threonine kinase family member 4), also known as KIAA0303, is a 2,626 amino acid protein that localizes to the cytoplasm and contains one protein kinase domain, as well as one PDZ domain and one AGC-kinase C-terminal domain. Expressed at high levels in most adult tissues, including testis, colon and small intestine, MAST4 uses magnesium as a cofactor to catalyze the ATP-dependent phosphorylation of target proteins. Multiple isoforms of MAST4 exist due to alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA0303; Microtubule-associated serine/threonine-protein kinase 4; PKC; unnamed protein product
Gene Aliases: KIAA0303; MAST4
UniProt ID: (Human) O15021
Entrez Gene ID: (Human) 375449
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support