Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MLAVPNMRFG IFLLWWGWVL ATESRMHWPG REVHEMSKKG RPQRQRREVH EDAHKQVSPI LRRSPDITKS PLTKSEQLLR; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels; however, they are structurally and functionally distinct. GABAC receptors (GABAC Rr) mediate rapid inhibitory neurotransmission in retina. Three human genes, r1 (GABRR1), r2 (GABRR2) and r3 (GABRR3), encode the three polypeptides that comprise this receptor. GABRR1 and GABRR2 are located close together, in a region of chromosome 6q that contains loci for inherited disorders of the eye, but GABRR3 maps to chromosome 3q11-q13.3. The r polypeptide genes, which are thought to share a common ancestor with GABA(A) receptor subunit genes, diverged at an early stage in the evolution of this gene family. The expression of GABAC Rr subunits is not restricted to the retina, but significant expression can also be detected in many other brain regions, especially in those belonging to the visual pathways.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA135P14.1 (gamma-aminobutyric acid (GABA) receptor, rho 1); GABA(A) receptor subunit rho-1; GABA(A) receptor, rho 1; GABA(C) receptor; GABA(C) rho1; GABAAR subunit rho-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho-1; gamma-aminobutyric acid (GABA) receptor, rho 1; gamma-aminobutyric acid (GABA-A) receptor subunit rho 1; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 1; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 1; gamma-aminobutyric acid A receptor, rho 1; Gamma-aminobutyric acid receptor subunit rho-1; gamma-aminobutyric acid type A receptor rho1 subunit; unnamed protein product
Gene Aliases: GABA-C; GABRR1
UniProt ID: (Human) P24046, (Mouse) P56475, (Rat) P50572
Entrez Gene ID: (Human) 2569, (Mouse) 14408, (Rat) 29694
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support