Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EZH2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001203247.1 | NP_001190176.1 | - | - | - |
NM_001203248.1 | NP_001190177.1 | - | - | - | |
NM_001203249.1 | NP_001190178.1 | - | - | - | |
NM_004456.4 | NP_004447.2 | 87277 | c.2081A>G | p.H694R | |
1087039 | c.2033T>G | p.F678C | |||
52980 | c.2069G>A | p.R690H | |||
220386 | c.2045C>G | p.A682G | |||
1315851 | c.2045C>T | p.A682V | |||
5020474 | c.2110+6T>G | p.? | |||
306072 | c.2051G>A | p.R684H | |||
6023078 | c.2051G>T | p.R684L | |||
53005 | c.2050C>T | p.R684C | |||
53040 | c.2080C>T | p.H694Y | |||
53009 | c.2079T>A | p.N693K | |||
6495159 | c.2054A>T | p.K685M | |||
1166639 | c.2083T>C | p.S695P | |||
4384248 | c.2048C>T | p.T683I | |||
600065 | c.2050C>A | p.R684S | |||
7409850 | c.2068C>A | p.R690S | |||
52981 | c.2078A>C | p.N693T | |||
6957422 | c.2064A>C | p.K688N | |||
220529 | c.2075C>T | p.A692V | |||
133038 | c.2035G>A | p.V679M | |||
3366842 | c.2030-1G>A | p.? | |||
6657560 | c.2089A>G | p.N697D | |||
7409581 | c.2110+1G>A | p.? | |||
5872165 | c.2038G>A | p.V680M | |||
1087037 | c.2085G>A | p.S695S | |||
49154 | c.2068C>T | p.R690C | |||
6495157 | c.2079T>G | p.N693K | |||
133040 | c.2110G>C | p.V704L | |||
4384246 | c.2084C>T | p.S695L | |||
53000 | c.2054_2054delA | p.K685fs*12 | |||
53033 | c.2078A>T | p.N693I | |||
NM_152998.2 | NP_694543.1 | 4384250 | c.1903G>A | p.V635M | |
5872166 | c.1906G>A | p.V636M | |||
306073 | c.1919G>A | p.R640H | |||
5020475 | c.1978+6T>G | p.? | |||
3366843 | c.1898-1G>A | p.? | |||
6495158 | c.1947T>G | p.N649K | |||
4384247 | c.1952C>T | p.S651L | |||
220530 | c.1943C>T | p.A648V | |||
6957423 | c.1932A>C | p.K644N | |||
7000500 | c.1918C>T | p.R640C | |||
1166640 | c.1951T>C | p.S651P | |||
1087040 | c.1901T>G | p.F634C | |||
3259625 | c.1936C>T | p.R646C | |||
4771583 | c.1949A>G | p.H650R | |||
6657561 | c.1957A>G | p.N653D | |||
1319003 | c.1937G>A | p.R646H | |||
1087038 | c.1953G>A | p.S651S | |||
6495160 | c.1922A>T | p.K641M | |||
220387 | c.1913C>G | p.A638G | |||
1234980 | c.1947T>A | p.N649K | |||
4384249 | c.1916C>T | p.T639I | |||
600066 | c.1918C>A | p.R640S | |||
XM_005249962.4 | XP_005250019.1 | - | - | - | |
XM_005249963.4 | XP_005250020.1 | - | - | - | |
XM_005249964.4 | XP_005250021.1 | - | - | - | |
XM_011515883.2 | XP_011514185.1 | - | - | - | |
XM_011515884.2 | XP_011514186.1 | - | - | - | |
XM_011515885.2 | XP_011514187.1 | - | - | - | |
XM_011515886.2 | XP_011514188.1 | - | - | - | |
XM_011515887.2 | XP_011514189.1 | - | - | - | |
XM_011515888.2 | XP_011514190.1 | - | - | - | |
XM_011515889.2 | XP_011514191.1 | - | - | - | |
XM_011515890.2 | XP_011514192.1 | - | - | - | |
XM_011515891.2 | XP_011514193.1 | - | - | - | |
XM_011515892.2 | XP_011514194.1 | - | - | - | |
XM_011515893.2 | XP_011514195.1 | - | - | - | |
XM_011515894.2 | XP_011514196.1 | - | - | - | |
XM_011515895.2 | XP_011514197.1 | - | - | - | |
XM_011515896.2 | XP_011514198.1 | - | - | - | |
XM_011515897.2 | XP_011514199.1 | - | - | - | |
XM_011515898.2 | XP_011514200.1 | - | - | - | |
XM_017011817.1 | XP_016867306.1 | - | - | - | |
XM_017011818.1 | XP_016867307.1 | - | - | - | |
XM_017011819.1 | XP_016867308.1 | - | - | - | |
XM_017011820.1 | XP_016867309.1 | - | - | - | |
XM_017011821.1 | XP_016867310.1 | - | - | - | |
GenBank | AB208895.1 | BAD92132.1 | - | - | - |
AK092676.1 | - | - | - | ||
AK293239.1 | - | - | - | ||
AK302216.1 | - | - | - | ||
AK314291.1 | - | - | - | ||
BC010858.2 | AAH10858.1 | - | - | - | |
BM974565.1 | - | - | - | ||
BU953788.1 | - | - | - | ||
ENST00000320356.0 | - | - | - | ||
JF327843.1 | - | - | - | ||
U61145.1 | AAC51520.1 | - | - | - | |
X95653.1 | CAA64955.1 | - | - | - |
Literature Links: |
EZH2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |