Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BIRC5 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001012270.1 | NP_001012270.1 | - | - | - |
NM_001012271.1 | NP_001012271.1 | 4843408 | c.399G>C | p.K133N | |
7156033 | c.349G>A | p.E117K | |||
3989371 | c.352G>A | p.E118K | |||
7205933 | c.323_324insGGTTGCG | p.F109fs*11 | |||
6002779 | c.393A>C | p.R131S | |||
3522793 | c.327C>T | p.F109F | |||
2745731 | c.321C>T | p.C107C | |||
5626714 | c.406A>G | p.I136V | |||
NM_001168.2 | NP_001159.2 | 4843409 | c.330G>C | p.K110N | |
3989372 | c.283G>A | p.E95K | |||
7205934 | c.254_255insGGTTGCG | p.F86fs*11 | |||
2745732 | c.252C>T | p.C84C | |||
7156034 | c.280G>A | p.E94K | |||
6002780 | c.324A>C | p.R108S | |||
3522794 | c.258C>T | p.F86F | |||
5626715 | c.337A>G | p.I113V | |||
GenBank | AB028869.1 | BAA93676.1 | - | - | - |
AB154416.1 | BAD11155.1 | - | - | - | |
AF077350.1 | AAD34226.1 | - | - | - | |
AK223428.1 | BAD97148.1 | - | - | - | |
AK301793.1 | - | - | - | ||
AK311917.1 | - | - | - | ||
AL547142.3 | - | - | - | ||
BC000784.2 | - | - | - | ||
BC008718.2 | AAH08718.1 | - | - | - | |
BC034148.1 | AAH34148.1 | - | - | - | |
BC065497.1 | AAH65497.1 | - | - | - | |
BE537971.1 | - | - | - | ||
BG178974.1 | - | - | - | ||
BI520186.1 | - | - | - | ||
BQ687404.1 | - | - | - | ||
CB112371.1 | - | - | - | ||
CR541740.1 | CAG46540.1 | - | - | - | |
DQ227257.1 | - | - | - | ||
DQ310375.1 | ABC42341.1 | - | - | - | |
DQ310376.1 | ABC42342.1 | - | - | - | |
DQ310377.1 | ABC42343.1 | - | - | - | |
DQ310378.1 | ABC42344.1 | - | - | - | |
DQ310379.1 | ABC42345.1 | - | - | - | |
DQ508245.1 | - | - | - | ||
DQ508246.1 | - | - | - | ||
DQ508247.1 | - | - | - | ||
DQ508248.1 | - | - | - | ||
DQ508249.1 | - | - | - | ||
ENST00000350051.0 | - | - | - | ||
HM625836.1 | - | - | - |
Literature Links: |
BIRC5 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |