Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DLG2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001142699.1 | NP_001136171.1 | 3791985 | c.2800G>A | p.E934K |
1586035 | c.2809G>T | p.E937* | |||
1146803 | c.2776G>A | p.D926N | |||
6644708 | c.2743C>T | p.R915C | |||
6070546 | c.2779C>G | p.R927G | |||
4920014 | c.2804T>C | p.F935S | |||
3870317 | c.2755G>A | p.E919K | |||
4168809 | c.2821+10C>T | p.? | |||
5458300 | c.2782G>T | p.A928S | |||
3383880 | c.2811A>G | p.E937E | |||
1586032 | c.2800G>T | p.E934* | |||
4970131 | c.2730-1G>A | p.? | |||
NM_001142700.1 | NP_001136172.1 | 5458303 | c.2104G>T | p.A702S | |
3383883 | c.2133A>G | p.E711E | |||
1146805 | c.2098G>A | p.D700N | |||
4970134 | c.2052-1G>A | p.? | |||
4168812 | c.2143+10C>T | p.? | |||
4920017 | c.2126T>C | p.F709S | |||
1586034 | c.2122G>T | p.E708* | |||
3870320 | c.2077G>A | p.E693K | |||
6070549 | c.2101C>G | p.R701G | |||
3791988 | c.2122G>A | p.E708K | |||
6644711 | c.2065C>T | p.R689C | |||
1586037 | c.2131G>T | p.E711* | |||
NM_001142702.1 | NP_001136174.1 | - | - | - | |
NM_001206769.1 | NP_001193698.1 | 1146804 | c.2578G>A | p.D860N | |
1586036 | c.2611G>T | p.E871* | |||
3383881 | c.2613A>G | p.E871E | |||
6070547 | c.2581C>G | p.R861G | |||
5458301 | c.2584G>T | p.A862S | |||
6644709 | c.2545C>T | p.R849C | |||
3870318 | c.2557G>A | p.E853K | |||
4970132 | c.2532-1G>A | p.? | |||
1586033 | c.2602G>T | p.E868* | |||
3791986 | c.2602G>A | p.E868K | |||
4920015 | c.2606T>C | p.F869S | |||
4168810 | c.2623+10C>T | p.? | |||
NM_001300983.1 | NP_001287912.1 | - | - | - | |
NM_001364.3 | NP_001355.2 | 932565 | c.2494G>T | p.E832* | |
3870319 | c.2440G>A | p.E814K | |||
4168811 | c.2506+10C>T | p.? | |||
4970133 | c.2415-1G>A | p.? | |||
4920016 | c.2489T>C | p.F830S | |||
5458302 | c.2467G>T | p.A823S | |||
690445 | c.2461G>A | p.D821N | |||
932566 | c.2485G>T | p.E829* | |||
3383882 | c.2496A>G | p.E832E | |||
3791987 | c.2485G>A | p.E829K | |||
6644710 | c.2428C>T | p.R810C | |||
6070548 | c.2464C>G | p.R822G | |||
XM_005273810.4 | XP_005273867.1 | - | - | - | |
XM_005273811.4 | XP_005273868.1 | - | - | - | |
XM_011544778.2 | XP_011543080.1 | - | - | - | |
XM_011544780.2 | XP_011543082.1 | - | - | - | |
XM_011544782.2 | XP_011543084.1 | - | - | - | |
XM_011544788.2 | XP_011543090.1 | - | - | - | |
XM_017017254.1 | XP_016872743.1 | - | - | - | |
XM_017017255.1 | XP_016872744.1 | - | - | - | |
XM_017017256.1 | XP_016872745.1 | - | - | - | |
XM_017017257.1 | XP_016872746.1 | - | - | - | |
XM_017017258.1 | XP_016872747.1 | - | - | - | |
XM_017017259.1 | XP_016872748.1 | - | - | - | |
XM_017017260.1 | XP_016872749.1 | - | - | - | |
XM_017017261.1 | XP_016872750.1 | - | - | - | |
XM_017017262.1 | XP_016872751.1 | - | - | - | |
XM_017017263.1 | XP_016872752.1 | - | - | - | |
XM_017017264.1 | XP_016872753.1 | - | - | - | |
XM_017017265.1 | XP_016872754.1 | - | - | - | |
XM_017017266.1 | XP_016872755.1 | - | - | - | |
XM_017017267.1 | XP_016872756.1 | - | - | - | |
XM_017017268.1 | XP_016872757.1 | - | - | - | |
XM_017017269.1 | XP_016872758.1 | - | - | - | |
XM_017017270.1 | XP_016872759.1 | - | - | - | |
XM_017017271.1 | XP_016872760.1 | - | - | - | |
XM_017017272.1 | XP_016872761.1 | - | - | - | |
XM_017017273.1 | XP_016872762.1 | - | - | - | |
XM_017017274.1 | XP_016872763.1 | - | - | - | |
XM_017017275.1 | XP_016872764.1 | - | - | - | |
XM_017017276.1 | XP_016872765.1 | - | - | - | |
XM_017017277.1 | XP_016872766.1 | - | - | - | |
XM_017017278.1 | XP_016872767.1 | - | - | - | |
XM_017017279.1 | XP_016872768.1 | - | - | - | |
XM_017017280.1 | XP_016872769.1 | - | - | - | |
XM_017017281.1 | XP_016872770.1 | - | - | - | |
XM_017017282.1 | XP_016872771.1 | - | - | - | |
XM_017017283.1 | XP_016872772.1 | - | - | - | |
XM_017017284.1 | XP_016872773.1 | - | - | - | |
XM_017017285.1 | XP_016872774.1 | - | - | - | |
XM_017017286.1 | XP_016872775.1 | - | - | - | |
XM_017017287.1 | XP_016872776.1 | - | - | - | |
XM_017017288.1 | XP_016872777.1 | - | - | - | |
XM_017017289.1 | XP_016872778.1 | - | - | - | |
XM_017017290.1 | XP_016872779.1 | - | - | - | |
XM_017017291.1 | XP_016872780.1 | - | - | - | |
XM_017017292.1 | XP_016872781.1 | - | - | - | |
GenBank | AK126776.1 | BAC86685.1 | - | - | - |
AK294375.1 | - | - | - | ||
AK295094.1 | - | - | - | ||
AK295624.1 | - | - | - | ||
AK299778.1 | - | - | - | ||
AK299797.1 | - | - | - | ||
BC143964.1 | - | - | - | ||
CR749820.1 | CAH18680.1 | - | - | - | |
CR933674.1 | CAI45970.1 | - | - | - | |
ENST00000398309.0 | - | - | - | ||
KJ535093.1 | - | - | - | ||
U32376.1 | AAB04949.1 | - | - | - |
Literature Links: |
DLG2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |