Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@1f7d949b
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Order Status
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
PIK3R1
Entrez Gene ID
5295
Gene Name:
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
Gene Chromosome Location
Chr.5: on Build GRCh38
UniGene
Hs.132225
Species:
Human
Primer Chromosome Location
Chr.5: 68226785-68227048 on Build GRCh38
View in UCSC browser
Amplicon Length
264
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00730567_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
PIK3R1
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_181523.2
NP_852664.1
5752841
c.196_197insG
p.D68fs*38
43188
c.211G>T
p.G71*
1438265
c.237_238insA
p.I82fs*24
6008944
c.253C>T
p.P85S
1438264
c.190delG
p.E65fs*10
7429196
c.150T>C
p.P50P
366838
c.176A>G
p.Y59C
4747412
c.238delA
p.I82fs*32
7346048
c.284delT
p.P96fs*18
5490676
c.219C>A
p.Y73*
5802333
c.197_198insG
p.D68fs*38
5487407
c.245_246insGGCC
p.I82fs*25
4714456
c.217_218insA
p.Y73fs*1
3617610
c.266C>T
p.P89L
6746549
c.279G>A
p.R93R
6946906
c.296_297insAG
p.G100fs*15
265734
c.271_272insCA
p.P92fs*23
194619
c.154G>T
p.E52*
4407349
c.219C>T
p.Y73Y
7344907
c.197G>A
p.R66K
4847727
c.325G>A
p.E109K
7351399
c.136C>T
p.Q46*
6588411
c.189A>C
p.T63T
5760378
c.251C>T
p.P84L
XM_005248542.3
XP_005248599.1
-
-
-
XM_017009585.1
XP_016865074.1
-
-
-
GenBank
BC094795.1
AAH94795.1
-
-
-
CT003423.1
-
-
-
ENST00000521381.0
-
-
-
M61906.1
-
-
-
Phenotype
Literature Links:
PIK3R1
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
enzyme modulator
kinase modulator
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-green:8080/10.253.224.144:8080.
git-commit: 512ede155cec2adea7292e762913190485b8e4d5
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/1.44.1-2022.06.24-1.0