Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10008 |
Gene Name: | potassium voltage-gated channel subfamily E regulatory subunit 3 |
Gene Aliases: |
HOKPP, HYPP, MiRP2 |
Location: |
Chr.11:74454841-74467729 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | 3 | 666 | NP_005463.1 |
XM_011544713.2 | 2 | 516 | XP_011543015.1 | |
XM_017017047.1 | 4 | 736 | XP_016872536.1 | |
XM_017017048.1 | 5 | 832 | XP_016872537.1 | |
XM_017017049.1 | 3 | 682 | XP_016872538.1 | |
XM_017017050.1 | 3 | 3540 | XP_016872539.1 | |
XM_017017051.1 | 4 | 655 | XP_016872540.1 | |
XM_017017052.1 | 3 | 606 | XP_016872541.1 | |
AF076531.1 | 2 | 411 | AAD28089.1 | |
AF302494.1 | 2 | 624 | AAG16255.1 | |
AK311876.1 | ||||
BC110612.2 | 2 | 435 | AAI10613.1 | |
BC113743.1 | 2 | 490 | AAI13744.1 | |
CR457209.1 | CAG33490.1 | |||
CR936625.1 | 3 | 660 | CAI56768.1 | |
DQ192291.1 | ABB00304.1 | |||
DQ192292.1 | ABB00305.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041154 | Chr.11:74403756 - 74466864 on Build GRCh38 | Loss | KCNE3 |
esv34979 | Chr.11:74167643 - 74822407 on Build GRCh38 | Gain | PGM2L1 POLD3 LOC100287896 RNF169 MIR4696 PPME1 KCNE3 CHRDL2 P4HA3 C2CD3 LOC101928580 LIPT2 MIR548AL |
esv3626840 | Chr.11:74371082 - 74464389 on Build GRCh38 | Gain | PGM2L1 MIR548AL KCNE3 |
esv2759837 | Chr.11:74131564 - 74846552 on Build GRCh38 | Gain | PGM2L1 POLD3 LOC100287896 RNF169 MIR4696 PPME1 KCNE3 CHRDL2 P4HA3 XRRA1 C2CD3 LOC101928580 LIPT2 MIR548AL |
nsv555443 | Chr.11:74398121 - 74469444 on Build GRCh38 | Loss | PGM2L1 MIR548AL KCNE3 |
nsv832209 | Chr.11:74444838 - 74618074 on Build GRCh38 | Gain | POLD3 LOC100287896 LIPT2 KCNE3 |
dgv209e214 | Chr.11:74225600 - 74828943 on Build GRCh38 | Gain | PGM2L1 POLD3 P4HA3 LOC100287896 RNF169 LOC101928580 LIPT2 MIR4696 MIR548AL PPME1 KCNE3 CHRDL2 |
nsv393 | Chr.11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr.11:74225622 - 74823410 on Build GRCh38 | Gain | PGM2L1 POLD3 P4HA3 LOC100287896 RNF169 LOC101928580 LIPT2 MIR4696 MIR548AL PPME1 KCNE3 CHRDL2 |
Set Membership: |
Intragenic Exonic DGV Variation |