Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4322 |
Gene Name: | matrix metallopeptidase 13 |
Gene Aliases: |
CLG3, MANDP1, MMP-13 |
Location: |
Chr.11:102942992-102955734 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MMP13 | NM_002427.3 | 10 | 1996 | NP_002418.1 |
CA413743.1 | ||||
X75308.1 | 10 | 1971 | CAA53056.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464 | Chr.11:102922044 - 102954570 on Build GRCh38 | Insertion | MMP13 |
nsv465 | Chr.11:102926638 - 102971487 on Build GRCh38 | Deletion | MMP13 |
Set Membership: |
Intragenic Exonic DGV Variation |