Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | 5 | 992 | NP_001304863.1 |
NM_001317935.1 | 5 | 980 | NP_001304864.1 | |
NM_020894.3 | 5 | 1093 | NP_065945.2 | |
XM_017008490.1 | 5 | 814 | XP_016863979.1 | |
XM_017008491.1 | 5 | 906 | XP_016863980.1 | |
XM_017008492.1 | 5 | 904 | XP_016863981.1 | |
XM_017008493.1 | 5 | 780 | XP_016863982.1 | |
XM_017008494.1 | 5 | 665 | XP_016863983.1 | |
XM_017008495.1 | 2 | 807 | XP_016863984.1 | |
XM_017008496.1 | 5 | 1417 | XP_016863985.1 | |
XM_017008497.1 | 5 | 1416 | XP_016863986.1 | |
XM_017008498.1 | 4 | 648 | XP_016863987.1 | |
XM_017008499.1 | 5 | 921 | XP_016863988.1 | |
AB040963.1 | 5 | 1093 | ||
BC110331.1 | 5 | 992 | AAI10332.1 | |
BC140901.1 | 5 | 830 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | CRIPAK PCGF3 LOC100130872 CTBP1-AS2 ATP5I IDUA LOC105374344 MFSD7 CTBP1-AS FGFRL1 SLC26A1 LOC101928548 LOC101928521 PDE6B MAEA TMEM175 NKX1-1 GAK MYL5 CTBP1 CPLX1 RNF212 DGKQ SPON2 LOC100129917 UVSSA LOC107986211 TMED11P |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | CRIPAK CTBP1 LOC101928548 SPON2 MAEA LOC100130872 UVSSA NKX1-1 CTBP1-AS2 CTBP1-AS |
nsv593301 | Chr.4:1306289 - 1358897 on Build GRCh38 | Loss | MAEA UVSSA |
nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss | CRIPAK UVSSA NKX1-1 |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | CTBP1 LOC101928548 SPON2 MAEA LOC100130872 UVSSA CTBP1-AS2 CTBP1-AS |
nsv593281 | Chr.4:1209477 - 1358897 on Build GRCh38 | Loss | CTBP1 MAEA UVSSA CTBP1-AS2 CTBP1-AS |
Set Membership: |
Intragenic Exonic DGV Variation |