Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4125 |
Gene Name: | mannosidase alpha class 2B member 1 |
Gene Aliases: |
LAMAN, MANB |
Location: |
Chr.19:12646508-12666777 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAN2B1 | NM_000528.3 | 6 | 954 | NP_000519.2 |
NM_001173498.1 | 6 | 954 | NP_001166969.1 | |
XM_005259913.1 | 6 | 932 | XP_005259970.1 | |
XM_017026818.1 | 4 | 616 | XP_016882307.1 | |
AB209921.1 | 6 | 866 | BAD93158.1 | |
AK291572.1 | 6 | 919 | ||
AK303418.1 | 7 | 893 | ||
AW172353.1 | 3 | 319 | ||
BC000736.2 | 6 | 900 | AAH00736.1 | |
U05572.1 | 6 | 911 | AAB03816.1 | |
U60266.1 | 6 | 892 | AAC34130.1 | |
U68382.1 | 6 | 859 | AAC50811.1 | |
U68567.1 | 6 | 866 | AAC50812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication | ASNA1 MIR5684 C19orf43 WDR83OS WDR83 TNPO2 DHPS SNORD135 ZNF791 LOC105372280 FBXW9 SNORD41 ZNF490 JUNB MAN2B1 HOOK2 RNASEH2A BEST2 PRDX2 |
dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion | ASNA1 MIR5684 C19orf43 WDR83OS WDR83 TNPO2 DHPS SNORD135 ZNF791 LOC105372280 FBXW9 SNORD41 JUNB MAN2B1 HOOK2 RNASEH2A BEST2 PRDX2 |
Set Membership: |
Intragenic Exonic DGV Variation |