Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 89781 |
Gene Name: | HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 |
Gene Aliases: |
BLOC3S2, LE |
Location: |
Chr.22:26443613-26483863 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HPS4 | NM_022081.5 | 14 | 4115 | NP_071364.4 |
NM_152841.2 | 12 | 3732 | NP_690054.1 | |
NR_073135.1 | 14 | 4133 | ||
NR_073136.1 | 13 | 3895 | ||
XM_011530485.1 | 16 | 4238 | XP_011528787.1 | |
XM_011530492.1 | XP_011528794.1 | |||
XM_011530493.2 | 14 | 3996 | XP_011528795.1 | |
XM_017029042.1 | 14 | 4011 | XP_016884531.1 | |
XM_017029043.1 | 15 | 4216 | XP_016884532.1 | |
XM_017029044.1 | 14 | 4014 | XP_016884533.1 | |
XM_017029045.1 | XP_016884534.1 | |||
XM_017029047.1 | 13 | 3942 | XP_016884536.1 | |
XM_017029048.1 | XP_016884537.1 | |||
XM_017029049.1 | XP_016884538.1 | |||
XM_017029050.1 | 12 | 3864 | XP_016884539.1 | |
XM_017029051.1 | 12 | 3769 | XP_016884540.1 | |
XM_017029052.1 | 15 | 4269 | XP_016884541.1 | |
AK097529.1 | 2 | 1520 | ||
BM975247.1 | 1 | 451 |
Set Membership: |
Intragenic Exonic non-DGV Variation |