Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55737 |
Gene Name: | VPS35 retromer complex component |
Gene Aliases: |
MEM3, PARK17 |
Location: |
Chr.16:46657979-46689232 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS35 | NM_018206.5 | 17 | 2964 | NP_060676.2 |
XM_005256045.3 | 16 | 2849 | XP_005256102.1 | |
AF186382.1 | 18 | 2883 | AAG40619.1 | |
BM971213.1 | 1 | 335 | ||
BQ574179.1 | 1 | 335 | ||
BX647213.1 | 10 | 4701 | CAI46268.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526656 | Chr.16:46505480 - 46812751 on Build GRCh38 | Gain | ANKRD26P1 ORC6 SHCBP1 VPS35 MYLK3 C16orf87 |
nsv428327 | Chr.16:46523056 - 46684578 on Build GRCh38 | Gain | ANKRD26P1 SHCBP1 VPS35 |
dgv2975n100 | Chr.16:46429858 - 46768065 on Build GRCh38 | Gain | ANKRD26P1 ORC6 SHCBP1 VPS35 MYLK3 |
esv3571142 | Chr.16:46430577 - 46840772 on Build GRCh38 | Gain | ANKRD26P1 ORC6 SHCBP1 VPS35 MYLK3 C16orf87 |
esv3638534 | Chr.16:46547097 - 46716009 on Build GRCh38 | Gain | ANKRD26P1 ORC6 SHCBP1 VPS35 MYLK3 |
Set Membership: |
Intragenic Exonic DGV Variation |