Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8518 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein |
Gene Aliases: |
DYS, ELP1, FD, IKAP, IKI3, TOT1 |
Location: |
Chr.9:108867520-108934332 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IKBKAP | NM_001318360.1 | 37 | 5923 | NP_001305289.1 |
NM_003640.4 | 37 | 6068 | NP_003631.2 | |
AF153419.2 | 37 | 5854 | AAG43369.1 | |
BF968300.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6659 | Chr.9:108856263 - 108890282 on Build GRCh38 | Insertion | ACTL7A IKBKAP |
Set Membership: |
Intragenic Exonic DGV Variation |