Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 90167 |
Gene Name: | FERM domain containing 7 |
Gene Aliases: |
NYS, NYS1, XIPAN |
Location: |
Chr.X:132076986-132128022 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRMD7 | NM_001306193.1 | NP_001293122.1 | ||
NM_194277.2 | NP_919253.1 | |||
XM_017029947.1 | XP_016885436.1 | |||
XM_017029948.1 | XP_016885437.1 | |||
XM_017029949.1 | XP_016885438.1 | |||
AK125336.1 | BAC86135.1 | |||
AL161984.1 | ||||
BC114371.1 | ||||
FJ717411.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |