Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 291 |
Gene Name: | solute carrier family 25 member 4 |
Gene Aliases: |
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1 |
Location: |
Chr.4:185143263-185150384 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A4 | NM_001151.3 | NP_001142.2 | ||
AB209764.1 | 1 | 1922 | BAD93001.1 | |
AK291902.1 | ||||
BC008664.1 | AAH08664.1 | |||
BC061589.1 | AAH61589.1 | |||
BC063643.1 | AAH63643.1 | |||
BG707401.1 | ||||
BI544547.1 | ||||
J02966.1 | AAA61223.1 | |||
KU177900.1 | ||||
KU177901.1 | ||||
KU177902.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv596369 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss | SLC25A4 MIR4455 CFAP97 SNX25 HELT LOC101928877 |
Set Membership: |
Intragenic Non-exonic DGV Variation |