Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55135 |
Gene Name: | WD repeat containing antisense to TP53 |
Gene Aliases: |
DKCB3, TCAB1, WDR79 |
Location: |
Chr.17:7686071-7703502 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP53 | NM_001143990.1 | NP_001137462.1 | ||
NM_001143991.1 | NP_001137463.1 | |||
NM_001143992.1 | NP_001137464.1 | |||
NM_018081.2 | NP_060551.2 | |||
XM_011523952.2 | XP_011522254.1 | |||
XM_017024812.1 | XP_016880301.1 | |||
AK001247.1 | BAA91579.1 | |||
AK056669.1 | ||||
AK092370.1 | ||||
AK225273.1 | ||||
AK225411.1 | ||||
AK303246.1 | ||||
AY766322.1 | ||||
BC002336.2 | AAH02336.1 | |||
DQ431240.1 | ||||
DQ431241.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | LOC100996842 SAT2 TNFSF13 TMEM256-PLSCR3 TMEM95 NEURL4 ACAP1 SNORD10 C17orf74 TNFSF12 TNK1 DLG4 GABARAP ALOX12 SENP3-EIF4A1 YBX2 EIF5A ACADVL SLC16A11 FXR2 SLC35G6 CD68 DVL2 WRAP53 CHRNB1 FGF11 LOC107983988 KCTD11 ASGR2 SHBG GPS2 ASGR1 TP53 CTDNEP1 ALOX12-AS1 TMEM256 CLDN7 MIR497HG RNASEK SNORA67 SENP3 PHF23 ATP1B2 NLGN2 SLC16A13 PLSCR3 SPEM1 RNASEK-C17orf49 SOX15 MIR497 ELP5 SLC2A4 MIR195 TMEM102 CLEC10A MIR324 TNFSF12-TNFSF13 ZBTB4 C17orf49 EIF4A1 BCL6B SNORA48 POLR2A MPDU1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |