Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7157 |
Gene Name: | tumor protein p53 |
Gene Aliases: |
BCC7, LFS1, P53, TRP53 |
Location: |
Chr.17:7668402-7687550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Entrez Gene ID: | 55135 |
Gene Name: | WD repeat containing antisense to TP53 |
Gene Aliases: |
DKCB3, TCAB1, WDR79 |
Location: |
Chr.17:7686071-7703502 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP53 | NM_001143990.1 | NP_001137462.1 | ||
AK056669.1 | 1 | 708 | ||
DQ431241.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | SHBG ACADVL MIR497 MIR195 ASGR2 LOC100996842 SLC2A4 DVL2 RNASEK-C17orf49 CTDNEP1 FGF11 TMEM95 ZBTB4 ATP1B2 CLDN7 TNFSF12 TNFSF13 C17orf49 PLSCR3 ACAP1 YBX2 GABARAP MIR497HG TMEM256-PLSCR3 CHRNB1 TNK1 SLC16A13 SPEM1 SENP3-EIF4A1 SNORD10 BCL6B DLG4 NEURL4 SAT2 KCTD11 SLC16A11 ALOX12 EIF5A PHF23 SNORA67 SNORA48 TMEM256 NLGN2 ELP5 ALOX12-AS1 GPS2 EIF4A1 CLEC10A SENP3 FXR2 TMEM102 WRAP53 SLC35G6 C17orf74 TNFSF12-TNFSF13 MPDU1 SOX15 TP53 POLR2A MIR324 ASGR1 RNASEK LOC107983988 CD68 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |