Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8546 |
Gene Name: | adaptor related protein complex 3 beta 1 subunit |
Gene Aliases: |
ADTB3, ADTB3A, HPS, HPS2, PE |
Location: |
Chr.5:78002326-78294755 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP3B1 | NM_001271769.1 | 27 | 3795 | NP_001258698.1 |
NM_003664.4 | 27 | 3783 | NP_003655.3 | |
XM_005248618.3 | 28 | 3791 | XP_005248675.1 | |
XM_017010001.1 | 28 | 3571 | XP_016865490.1 | |
BC038444.1 | 27 | 3744 | AAH38444.1 | |
BX538041.1 | 27 | 3732 | CAD97982.1 | |
U81504.1 | 27 | 3699 | AAB61638.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |