Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1135 |
Gene Name: | cholinergic receptor nicotinic alpha 2 subunit |
Gene Aliases: |
- |
Location: |
Chr.8:27459761-27479296 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHRNA2 | NM_000742.3 | 6 | 1282 | NP_000733.2 |
NM_001282455.1 | 6 | 1237 | NP_001269384.1 | |
XM_005273397.2 | 6 | 1201 | XP_005273454.1 | |
XM_006716282.1 | 7 | 1193 | XP_006716345.1 | |
XM_011544388.1 | 7 | 1284 | XP_011542690.1 | |
XM_011544389.1 | 6 | 1146 | XP_011542691.1 | |
XM_017013003.1 | 6 | 1209 | XP_016868492.1 | |
XM_017013004.1 | 7 | 1012 | XP_016868493.1 | |
AK296348.1 | 6 | 792 | ||
AK308465.1 | ||||
AK314787.1 | 6 | 1067 | ||
BC153866.1 | 5 | 708 | ||
CD013901.1 | 5 | 848 | ||
U62431.1 | 6 | 1227 | AAB40109.1 | |
Y16281.1 | 5 | 673 | CAA76154.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1020582 | Chr.8:27452251 - 27549327 on Build GRCh38 | Gain | CHRNA2 EPHX2 PTK2B |
Set Membership: |
Intragenic Exonic DGV Variation |