Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 81494 |
Gene Name: | complement factor H related 5 |
Gene Aliases: |
CFHL5, CFHR5D, FHR-5, FHR5 |
Location: |
Chr.1:196975022-197009725 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CFHR5 | NM_030787.3 | 1 | 146 | NP_110414.1 |
XM_011510020.2 | XP_011508322.1 | |||
AF295327.1 | 1 | 111 | AAK15619.1 | |
AK314026.1 | 1 | 146 | ||
BC026282.1 | 1 | 127 | ||
BC111773.1 | 1 | 111 | AAI11774.1 | |
DA638244.1 | 1 | 146 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12e55 | Chr.1:196762018 - 197042447 on Build GRCh38 | Gain | CFHR2 F13B CFHR5 CFHR3 CFHR4 CFHR1 |
nsv469599 | Chr.1:196827346 - 196986100 on Build GRCh38 | Loss | CFHR2 CFHR5 CFHR4 CFHR1 |
nsv428290 | Chr.1:196621707 - 196992672 on Build GRCh38 | Gain+Loss | CFHR2 CFH CFHR5 CFHR3 CFHR4 CFHR1 |
esv3588486 | Chr.1:196970045 - 196983172 on Build GRCh38 | Loss | CFHR5 |
nsv471614 | Chr.1:196700381 - 196986100 on Build GRCh38 | Gain+Loss | CFHR2 CFH CFHR5 CFHR3 CFHR4 CFHR1 |
esv2721051 | Chr.1:196970051 - 196983169 on Build GRCh38 | Deletion | CFHR5 |
nsv528460 | Chr.1:196573437 - 197477572 on Build GRCh38 | Gain | CFHR2 CRB1 CFH ZBTB41 F13B MIR4735 CFHR5 CFHR3 KCNT2 CFHR4 ASPM CFHR1 |
nsv8702 | Chr.1:196974722 - 196985445 on Build GRCh38 | Loss | CFHR5 |
Set Membership: |
Intragenic Exonic DGV Variation |