Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 51314 |
Gene Name: | NME/NM23 family member 8 |
Gene Aliases: |
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2 |
Location: |
Chr.7:37848597-37900401 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NME8 | NM_016616.4 | NP_057700.3 | ||
AF202051.2 | AAF20909.2 | |||
AF305596.1 | AAN04258.1 | |||
AK313599.1 | ||||
BC036816.1 | AAH36816.1 | |||
GQ472221.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv24163 | Chr.7:37891422 - 37897099 on Build GRCh38 | Gain | NME8 |
esv3612864 | Chr.7:37891337 - 37897492 on Build GRCh38 | Gain | NME8 |
nsv8074 | Chr.7:37825738 - 38170485 on Build GRCh38 | Loss | NME8 EPDR1 SFRP4 |
esv2761091 | Chr.7:37891879 - 37896928 on Build GRCh38 | Gain | NME8 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |