Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 6392 |
Gene Name: | succinate dehydrogenase complex subunit D |
Gene Aliases: |
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS |
Location: |
Chr.11:112086824-112095801 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDHD | NM_001276503.1 | NP_001263432.1 | ||
NM_001276504.1 | NP_001263433.1 | |||
NM_001276506.1 | NP_001263435.1 | |||
NM_003002.3 | NP_002993.1 | |||
NR_077060.1 | ||||
AB006202.1 | BAA22054.1 | |||
AK075360.1 | ||||
BC005263.1 | AAH05263.1 | |||
BC009574.1 | AAH09574.1 | |||
BC012603.1 | AAH12603.1 | |||
BC015188.1 | AAH15188.1 | |||
BC015992.1 | AAH15992.1 | |||
BC022350.1 | AAH22350.1 | |||
BC070307.1 | AAH70307.1 | |||
BC071755.1 | AAH71755.1 | |||
BC071756.1 | AAH71756.1 | |||
BE567994.1 | ||||
BF211358.1 | ||||
BG492859.1 | ||||
BQ932668.1 | ||||
BT007238.1 | AAP35902.1 | |||
BX419391.2 | ||||
CD105572.1 | ||||
CR456932.1 | CAG33213.1 | |||
KU178325.1 | ||||
KU178326.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv975979 | Chr.11:112088953 - 112096507 on Build GRCh38 | Duplication | SDHD |
esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion | SDHD IL18 C11orf57 BCO2 DLAT PIH1D2 TEX12 DIXDC1 TIMM8B |
Set Membership: |
Intragenic Non-exonic DGV Variation |