Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 124590 |
Gene Name: | USH1 protein network component sans |
Gene Aliases: |
ANKS4A, SANS |
Location: |
Chr.17:74916083-74923263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USH1G | NM_001282489.2 | NP_001269418.1 | ||
NM_173477.4 | NP_775748.2 | |||
XM_011524296.2 | XP_011522598.1 | |||
AK091243.1 | BAC03619.1 | |||
AK289804.1 | ||||
AK296899.1 | ||||
BC101096.2 | AAI01097.1 | |||
BC101097.1 | AAI01098.1 | |||
BC101098.2 | AAI01099.1 | |||
BC101099.1 | AAI01100.1 | |||
DC403891.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss | GRIN2C OTOP3 MRPL58 OTOP2 CDR2L HID1 USH1G FADS6 TMEM104 FDXR HID1-AS1 |
nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss | GRIN2C OTOP3 OTOP2 HID1 USH1G FADS6 TMEM104 FDXR HID1-AS1 |
nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion | GRIN2C OTOP3 OTOP2 SLC9A3R1 NAT9 HID1 USH1G FADS6 TMEM104 FDXR |
nsv576026 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain | OTOP3 OTOP2 HID1 USH1G FADS6 HID1-AS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |