Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5889 |
Gene Name: | RAD51 paralog C |
Gene Aliases: |
BROVCA3, FANCO, R51H3, RAD51L2 |
Location: |
Chr.17:58692538-58734342 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51C | NM_058216.2 | NP_478123.1 | ||
NR_103872.1 | ||||
XM_006722001.3 | XP_006722064.1 | |||
XM_006722002.3 | XP_006722065.1 | |||
XM_006722004.3 | XP_006722067.1 | |||
XM_006722005.3 | XP_006722068.1 | |||
XM_011525092.2 | XP_011523394.1 | |||
XM_011525093.2 | XP_011523395.1 | |||
XM_011525094.2 | XP_011523396.1 | |||
XM_017024914.1 | XP_016880403.1 | |||
XM_017024915.1 | XP_016880404.1 | |||
XM_017024916.1 | XP_016880405.1 | |||
XM_017024917.1 | XP_016880406.1 | |||
XM_017024918.1 | XP_016880407.1 | |||
XM_017024919.1 | XP_016880408.1 | |||
AF029669.1 | AAC39604.1 | |||
AK290487.1 | ||||
AK303363.1 | ||||
BC093930.1 | AAH93930.1 | |||
BC101485.1 | AAI01486.1 | |||
BC107753.1 | AAI07754.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss | TEX14 PPM1E RAD51C |
nsv1066945 | Chr.17:58650965 - 58704810 on Build GRCh38 | Loss | TEX14 RAD51C |
nsv428347 | Chr.17:58565015 - 58717588 on Build GRCh38 | Gain | TEX14 RAD51C |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |