Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 29888 |
Gene Name: | striatin 4 |
Gene Aliases: |
PPP2R6C, ZIN, zinedin |
Location: |
Chr.19:46719509-46746475 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STRN4 | NM_001039877.1 | NP_001034966.1 | ||
NM_013403.2 | NP_037535.2 | |||
XM_006723171.2 | XP_006723234.1 | |||
XM_006723172.2 | XP_006723235.1 | |||
XM_011526878.1 | XP_011525180.1 | |||
XM_017026719.1 | XP_016882208.1 | |||
AB209003.1 | BAD92240.1 | |||
AF212940.1 | AAF29527.1 | |||
AK297637.1 | ||||
AK298804.1 | ||||
BC034604.1 | AAH34604.1 | |||
DA510367.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss | FKRP LOC107987269 MIR320E SNAR-E STRN4 PRKD2 AP2S1 SLC1A5 ARHGAP35 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |