Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9992 |
Gene Name: | potassium voltage-gated channel subfamily E regulatory subunit 2 |
Gene Aliases: |
ATFB4, LQT5, LQT6, MIRP1 |
Location: |
Chr.21:34364024-34371141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNE2 | NM_172201.1 | NP_751951.1 | ||
AF302095.1 | AAG13416.1 | |||
BC093892.1 | AAH93892.1 | |||
BC112087.1 | AAI12088.1 | |||
DQ192290.1 | ABB00303.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7824n54 | Chr.21:34352440 - 34535115 on Build GRCh38 | Gain | SMIM11A RCAN1 C21orf140 LOC105372793 KCNE1 LOC388820 KCNE2 |
dgv4426n100 | Chr.21:34339322 - 34534919 on Build GRCh38 | Gain | SMIM11A RCAN1 C21orf140 LOC105372793 KCNE1 LOC388820 KCNE2 |
nsv834088 | Chr.21:34241420 - 34379776 on Build GRCh38 | Loss | SMIM11A KCNE2 |
nsv1055429 | Chr.21:34299853 - 34516938 on Build GRCh38 | Gain | SMIM11A RCAN1 C21orf140 LOC105372793 KCNE1 LOC388820 KCNE2 |
esv3575386 | Chr.21:34353927 - 34534570 on Build GRCh38 | Gain | SMIM11A RCAN1 C21orf140 LOC105372793 KCNE1 LOC388820 KCNE2 |
esv2762109 | Chr.21:34350696 - 34531755 on Build GRCh38 | Gain | SMIM11A RCAN1 C21orf140 LOC105372793 KCNE1 LOC388820 KCNE2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |