Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8292 |
Gene Name: | collagen like tail subunit of asymmetric acetylcholinesterase |
Gene Aliases: |
CMS5, EAD |
Location: |
Chr.3:15450133-15521751 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COLQ | NM_005677.3 | NP_005668.2 | ||
NM_080538.2 | NP_536799.1 | |||
NM_080539.3 | NP_536800.2 | |||
AF057036.1 | AAC39927.1 | |||
AJ225895.1 | CAA12648.1 | |||
AK128401.1 | ||||
AK289688.1 | ||||
AY150334.1 | AAO06814.1 | |||
AY150335.1 | AAO06815.1 | |||
AY150336.1 | AAO06816.1 | |||
AY150337.1 | AAO06817.1 | |||
AY150338.1 | AAO06818.1 | |||
AY150339.1 | AAO06819.1 | |||
BC074828.2 | AAH74828.1 | |||
BC074829.2 | AAH74829.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834622 | Chr.3:15397807 - 15587083 on Build GRCh38 | Gain | METTL6 COLQ HACL1 MIR4270 EAF1 |
nsv834621 | Chr.3:15324275 - 15509456 on Build GRCh38 | Gain | METTL6 COLQ MIR4270 EAF1 SH3BP5 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |