Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | NP_001304863.1 | ||
NM_001317935.1 | NP_001304864.1 | |||
NM_020894.3 | NP_065945.2 | |||
XM_017008490.1 | XP_016863979.1 | |||
XM_017008491.1 | XP_016863980.1 | |||
XM_017008492.1 | XP_016863981.1 | |||
XM_017008493.1 | XP_016863982.1 | |||
XM_017008494.1 | XP_016863983.1 | |||
XM_017008495.1 | XP_016863984.1 | |||
XM_017008498.1 | XP_016863987.1 | |||
XM_017008500.1 | XP_016863989.1 | |||
AB040963.1 | ||||
AK292661.1 | ||||
BC110331.1 | AAI10332.1 | |||
BC140901.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | RNF212 SPON2 CRIPAK PDE6B IDUA GAK LOC100130872 NKX1-1 LOC105374344 UVSSA MYL5 ATP5I SLC26A1 LOC101928521 CTBP1-AS2 CTBP1-AS PCGF3 TMEM175 FGFRL1 CPLX1 LOC101928548 MFSD7 LOC100129917 LOC107986211 MAEA DGKQ TMED11P CTBP1 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | CTBP1-AS2 SPON2 CRIPAK UVSSA CTBP1-AS MAEA LOC101928548 LOC100130872 NKX1-1 CTBP1 |
nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss | CRIPAK UVSSA NKX1-1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |