Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 154881 |
Gene Name: | potassium channel tetramerization domain containing 7 |
Gene Aliases: |
CLN14, EPM3 |
Location: |
Chr.7:66628881-66643229 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD7 | NM_001167961.2 | 5 | 2951 | NP_001161433.1 |
NM_153033.4 | 4 | 4116 | NP_694578.1 | |
BU682845.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761151 | Chr.7:66630751 - 66673298 on Build GRCh38 | Gain | LOC100996437 KCTD7 |
esv3613515 | Chr.7:66635366 - 66657471 on Build GRCh38 | Gain | LOC100996437 KCTD7 |
esv3891135 | Chr.7:66376216 - 66777606 on Build GRCh38 | Gain | GS1-124K5.11 GS1-124K5.4 RABGEF1 LOC100996437 KCTD7 LINC00174 |
esv3613516 | Chr.7:66636982 - 66673774 on Build GRCh38 | Gain | LOC100996437 KCTD7 |
esv2665862 | Chr.7:66635706 - 66672765 on Build GRCh38 | Deletion | LOC100996437 KCTD7 |
Set Membership: |
Intragenic Exonic DGV Variation |