Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2990 |
Gene Name: | glucuronidase beta |
Gene Aliases: |
BG, MPS7 |
Location: |
Chr.7:65960684-65982314 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GUSB | NM_000181.3 | NP_000172.2 | ||
NM_001284290.1 | NP_001271219.1 | |||
NM_001293104.1 | NP_001280033.1 | |||
NM_001293105.1 | NP_001280034.1 | |||
NR_120531.1 | ||||
XM_005250297.3 | XP_005250354.1 | |||
XM_011516113.1 | XP_011514415.1 | |||
XM_011516114.1 | XP_011514416.1 | |||
XM_017012091.1 | XP_016867580.1 | |||
XM_017012092.1 | XP_016867581.1 | |||
XM_017012093.1 | XP_016867582.1 | |||
AA947121.1 | ||||
AK096764.1 | ||||
AK223406.1 | BAD97126.1 | |||
AK300223.1 | ||||
AK303819.1 | ||||
AK312752.1 | ||||
BC014142.2 | AAH14142.1 | |||
BM719712.1 | ||||
CD014092.1 | ||||
CD014093.1 | ||||
CD014094.1 | ||||
M15182.1 | AAA52561.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1028971 | Chr.7:65949077 - 65992450 on Build GRCh38 | Gain | GUSB VKORC1L1 |
esv3613492 | Chr.7:65946577 - 66001473 on Build GRCh38 | Gain | GUSB VKORC1L1 |
esv2761079 | Chr.7:65601260 - 66224746 on Build GRCh38 | Gain+Loss | INTS4P2 CCT6P1 SNORA22 CRCP GUSB ASL TPST1 VKORC1L1 LOC441242 LOC84214 |
esv3613493 | Chr.7:65949620 - 65975662 on Build GRCh38 | Gain | GUSB VKORC1L1 |
esv2664145 | Chr.7:65814760 - 66395282 on Build GRCh38 | Deletion | CRCP GUSB ASL TPST1 VKORC1L1 LOC84214 LINC00174 |
esv21527 | Chr.7:65948375 - 66003033 on Build GRCh38 | Gain | GUSB VKORC1L1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |