Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 54903 |
Gene Name: | Meckel syndrome, type 1 |
Gene Aliases: |
BBS13, MES, MKS, POC12 |
Location: |
Chr.17:58205436-58219605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MKS1 | NM_001165927.1 | NP_001159399.1 | ||
NM_001321268.1 | NP_001308197.1 | |||
NM_001321269.1 | NP_001308198.1 | |||
NM_017777.3 | NP_060247.2 | |||
XM_005257485.3 | XP_005257542.1 | |||
XM_006721965.2 | XP_006722028.1 | |||
XM_011524957.2 | XP_011523259.1 | |||
XM_011524958.2 | XP_011523260.1 | |||
XM_011524959.2 | XP_011523261.1 | |||
XM_011524960.2 | XP_011523262.1 | |||
XM_017024803.1 | XP_016880292.1 | |||
XM_017024804.1 | XP_016880293.1 | |||
XM_017024805.1 | XP_016880294.1 | |||
AK301020.1 | ||||
AK310815.1 | ||||
BC010061.2 | AAH10061.1 | |||
DQ185029.1 | AAZ94714.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517229 | Chr.17:58176226 - 58373163 on Build GRCh38 | Gain+Loss | TSPOAP1 LPO MIR142 MPO MKS1 RNF43 MIR4736 SUPT4H1 EPX TSPOAP1-AS1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |