Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | NP_001304863.1 | ||
NM_001317935.1 | NP_001304864.1 | |||
NM_020894.3 | NP_065945.2 | |||
XM_017008490.1 | XP_016863979.1 | |||
XM_017008491.1 | XP_016863980.1 | |||
XM_017008492.1 | XP_016863981.1 | |||
XM_017008493.1 | XP_016863982.1 | |||
XM_017008494.1 | XP_016863983.1 | |||
XM_017008495.1 | XP_016863984.1 | |||
XM_017008498.1 | XP_016863987.1 | |||
AB040963.1 | ||||
BC110331.1 | AAI10332.1 | |||
BC140901.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | LOC101928521 CPLX1 SLC26A1 LOC107986211 TMED11P DGKQ IDUA NKX1-1 MAEA FGFRL1 UVSSA GAK MFSD7 PCGF3 LOC101928548 CRIPAK RNF212 LOC100130872 CTBP1-AS2 PDE6B CTBP1 CTBP1-AS MYL5 LOC105374344 LOC100129917 ATP5I TMEM175 SPON2 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | LOC101928548 CTBP1-AS NKX1-1 CRIPAK MAEA LOC100130872 UVSSA CTBP1-AS2 SPON2 CTBP1 |
nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss | NKX1-1 CRIPAK UVSSA |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |