Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1135 |
Gene Name: | cholinergic receptor nicotinic alpha 2 subunit |
Gene Aliases: |
- |
Location: |
Chr.8:27459761-27479296 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHRNA2 | NM_000742.3 | NP_000733.2 | ||
NM_001282455.1 | NP_001269384.1 | |||
XM_005273397.2 | XP_005273454.1 | |||
XM_006716282.1 | XP_006716345.1 | |||
XM_011544388.1 | XP_011542690.1 | |||
XM_011544389.1 | XP_011542691.1 | |||
XM_017013003.1 | XP_016868492.1 | |||
XM_017013004.1 | XP_016868493.1 | |||
AK296348.1 | ||||
AK314787.1 | ||||
DA407151.1 | ||||
U62431.1 | AAB40109.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831275 | Chr.8:27470773 - 27624088 on Build GRCh38 | Loss | MIR6843 CLU EPHX2 CHRNA2 |
nsv1020582 | Chr.8:27452251 - 27549327 on Build GRCh38 | Gain | EPHX2 PTK2B CHRNA2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |