Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10806 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Location: |
Chr.1:243255328-243500092 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833070 | Chr.1:243339809 - 243537501 on Build GRCh38 | Gain | MIR4677 AKT3 SDCCAG8 |
nsv1001488 | Chr.1:243227523 - 243419972 on Build GRCh38 | Loss | MIR4677 CEP170 SDCCAG8 |
nsv517939 | Chr.1:243213455 - 243359604 on Build GRCh38 | Gain | MIR4677 CEP170 SDCCAG8 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |