Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57091 |
Gene Name: | Cas scaffolding protein family member 4 |
Gene Aliases: |
C20orf32, CAS4, HEFL, HEPL |
Location: |
Chr.20:56411550-56460240 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASS4 | NM_001164114.1 | NP_001157586.1 | ||
NM_001164115.1 | NP_001157587.1 | |||
NM_001164116.1 | NP_001157588.1 | |||
NM_020356.3 | NP_065089.2 | |||
XM_006723831.3 | XP_006723894.1 | |||
AJ276678.1 | CAC00655.1 | |||
AK027760.1 | BAB55351.1 | |||
AK295613.1 | ||||
AK308057.1 | ||||
AK314653.1 | ||||
BC027951.1 | AAH27951.1 | |||
BX280195.1 | ||||
DA584687.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv787e214 | Chr.20:56429038 - 56481532 on Build GRCh38 | Gain | CASS4 RTFDC1 |
esv3646189 | Chr.20:56430399 - 56481532 on Build GRCh38 | Loss | CASS4 RTFDC1 |
nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain | CASS4 TFAP2C CSTF1 GCNT7 FAM209A LOC105372682 FAM209B RTFDC1 |
nsv834013 | Chr.20:56307279 - 56476126 on Build GRCh38 | Gain | CASS4 CSTF1 AURKA FAM210B RTFDC1 |
esv3646191 | Chr.20:56431805 - 56436322 on Build GRCh38 | Loss | CASS4 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |