Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 73750 |
Gene Name: | whirlin |
Gene Aliases: |
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi |
Location: |
Chr.4:63414909-63496078 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Whrn | NM_001008791.2 | NP_001008791.1 | ||
NM_001008792.2 | NP_001008792.1 | |||
NM_001008793.2 | NP_001008793.1 | |||
NM_028640.2 | NP_082916.1 | |||
AK122523.1 | BAC65805.1 | |||
AK157955.1 | BAE34281.1 | |||
AY739114.1 | AAV87519.1 | |||
AY739115.1 | AAV87520.1 | |||
AY739116.1 | AAV87521.1 | |||
AY739117.1 | AAV87522.1 | |||
AY739118.1 | AAV87523.1 | |||
AY739119.1 | AAV87524.1 | |||
AY739120.1 | AAV87525.1 | |||
AY739121.1 | AAV87526.1 | |||
AY739122.1 | AAV87527.1 | |||
BC150757.1 | ||||
HQ148552.1 |
Set Membership: |
Intragenic Non-exonic |