Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 16334 |
Gene Name: | insulin II |
Gene Aliases: |
AA986540, Ins-2, InsII, Mody, Mody4, proinsulin |
Location: |
Chr.7:142678656-142679708 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ins2 | NM_001185083.1 | NP_001172012.1 | ||
NM_001185084.1 | NP_001172013.1 | |||
NM_008387.4 | NP_032413.1 | |||
AK007612.1 | BAB25135.1 | |||
AY899305.1 | AAW83742.1 | |||
BC099934.1 | ||||
BC132650.1 | ||||
BC132652.1 | ||||
BC145554.1 | ||||
GQ915612.1 | ||||
Z47785.1 |
Set Membership: |
Intragenic Non-exonic |