Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 17863 |
Gene Name: | myeloblastosis oncogene |
Gene Aliases: |
AI550390, M16449, c-myb |
Location: |
Chr.10:21124930-21160984 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myb | NM_001198914.1 | 5 | 648 | NP_001185843.1 |
NM_010848.3 | 5 | 648 | NP_034978.3 | |
AK088079.1 | 5 | 569 | BAC40133.1 | |
AK088598.1 | 5 | 570 | BAC40443.1 | |
AK132250.1 | 5 | 513 | ||
AK137965.1 | 5 | 514 | ||
BC011513.1 | 5 | 556 | AAH11513.1 | |
M12848.1 | 5 | 646 | AAB59713.1 | |
X02774.1 | 5 | 418 | CAA26551.1 |