Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 17863 |
Gene Name: | myeloblastosis oncogene |
Gene Aliases: |
AI550390, M16449, c-myb |
Location: |
Chr.10:21124930-21160984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myb | NM_001198914.1 | NP_001185843.1 | ||
NM_010848.3 | NP_034978.3 | |||
AK036518.1 | 1 | 811 | ||
AK088079.1 | BAC40133.1 | |||
AK088598.1 | BAC40443.1 | |||
AK132250.1 | ||||
AK137794.1 | BAE23498.1 | |||
AK137965.1 | ||||
BC011513.1 | AAH11513.1 | |||
CN693253.1 | ||||
M12848.1 | AAB59713.1 | |||
X02774.1 | CAA26551.1 |