Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9990 |
Gene Name: | solute carrier family 12 member 6 |
Gene Aliases: |
ACCPN, KCC3, KCC3A, KCC3B |
Location: |
Chr.15:34229996-34338064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC12A6 | NM_001042494.1 | NP_001035959.1 | ||
NM_001042495.1 | NP_001035960.1 | |||
NM_001042496.1 | NP_001035961.1 | |||
NM_001042497.1 | NP_001035962.1 | |||
NM_005135.2 | NP_005126.1 | |||
NM_133647.1 | NP_598408.1 | |||
XM_006720793.3 | XP_006720856.1 | |||
XM_011522269.2 | XP_011520571.1 | |||
AF105366.1 | AAD39742.1 | |||
AF108831.1 | AAD25337.1 | |||
AF116242.1 | AAF24986.1 | |||
AF477977.1 | AAL85335.1 | |||
AF531258.1 | AAQ10026.1 | |||
AF531259.1 | AAQ10027.1 | |||
AF531260.1 | AAQ10028.1 | |||
AK128133.1 | ||||
AK292550.1 | ||||
AK315283.1 | ||||
AL117500.1 | CAB55965.1 | |||
BC051744.1 | AAH51744.1 | |||
BC070107.1 | ||||
BC126241.1 | ||||
BC126243.1 | ||||
BX648195.1 | CAI46042.1 | |||
DQ138323.1 | ABA02873.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain | RYR3 ARHGAP11A FMN1 LPCAT4 LOC105370757 AVEN NUTM1 SCG5 MIR1233-2 CHRM5 EMC7 KATNBL1 GOLGA8B NOP10 LOC100131315 EMC4 TMCO5B MIR1233-1 SLC12A6 GREM1 LOC101928042 GOLGA8A PGBD4 LOC101928134 |
esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | RYR3 ARHGAP11A GOLGA8K LPCAT4 CHRNA7 AVEN GOLGA8N EMC7 GOLGA8B NOP10 EMC4 TMCO5B MIR1233-1 GOLGA8A LOC100996255 LOC100653133 WHAMMP1 LOC101928134 FMN1 LOC105370757 LOC101060588 NUTM1 LOC105370940 SCG5 MIR1233-2 CHRM5 KATNBL1 LOC100131315 SLC12A6 GREM1 LOC101928042 PGBD4 GOLGA8O ULK4P1 |
esv2760024 | Chr.15:34250820 - 34696046 on Build GRCh38 | Gain+Loss | GOLGA8B NOP10 LPCAT4 MIR1233-1 SLC12A6 NUTM1 GOLGA8A MIR1233-2 |
esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain | RYR3 LPCAT4 AVEN NUTM1 MIR1233-2 CHRM5 EMC7 KATNBL1 GOLGA8B NOP10 AQR LOC101928174 EMC4 MIR1233-1 SLC12A6 GOLGA8A PGBD4 GJD2 ZNF770 ACTC1 |
dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain | RYR3 ARHGAP11A LPCAT4 AVEN GOLGA8N EMC7 GOLGA8B NOP10 EMC4 TMCO5B MIR1233-1 GOLGA8A LOC101928134 FMN1 LOC105370757 NUTM1 SCG5 MIR1233-2 CHRM5 KATNBL1 LOC100131315 SLC12A6 GREM1 LOC101928042 PGBD4 |
Set Membership: |
Intragenic Non-exonic DGV Variation |