Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 6792 |
Gene Name: | cyclin dependent kinase like 5 |
Gene Aliases: |
CFAP247, EIEE2, ISSX, STK9 |
Location: |
Chr.X:18425605-18653629 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDKL5 | NM_001037343.1 | NP_001032420.1 | ||
NM_003159.2 | NP_003150.1 | |||
AY217744.1 | AAO64440.1 | |||
Y15057.1 | CAA75342.1 |
Entrez Gene ID: | 6247 |
Gene Name: | retinoschisin 1 |
Gene Aliases: |
RS, XLRS1 |
Location: |
Chr.X:18639688-18672103 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RS1 | NM_000330.3 | 6 | 1376 | NP_000321.1 |
AF014459.1 | 6 | 1356 | AAC17928.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6821 | Chr.X:18622224 - 18656388 on Build GRCh38 | Insertion | RS1 CDKL5 |
nsv528654 | Chr.X:18627439 - 18660570 on Build GRCh38 | Gain | RS1 CDKL5 |
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |