Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 54897 |
Gene Name: | castor zinc finger 1 |
Gene Aliases: |
CAS11, CST, SRG, ZNF693, dJ734G22.1 |
Location: |
Chr.1:10636602-10796676 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASZ1 | NM_001079843.2 | 21 | 7319 | NP_001073312.1 |
XM_017001539.1 | 20 | 7483 | XP_016857028.1 | |
XM_017001540.1 | 22 | 7593 | XP_016857029.1 | |
XM_017001541.1 | 22 | 7655 | XP_016857030.1 | |
AK130996.1 | 1 | 547 | BAC85474.1 | |
DQ217660.1 | 21 | 7291 | ||
DQ372703.1 | 1 | 1246 | ABD14411.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4n27 | Chr.1:10613137 - 10736809 on Build GRCh38 | Loss | PEX14 CASZ1 |
dgv105n54 | Chr.1:10613037 - 10736809 on Build GRCh38 | Loss | PEX14 CASZ1 |
nsv828141 | Chr.1:10635420 - 10644140 on Build GRCh38 | Gain | CASZ1 |
esv29966 | Chr.1:10616743 - 10814095 on Build GRCh38 | Loss | PEX14 CASZ1 |
nsv545400 | Chr.1:10624041 - 10781964 on Build GRCh38 | Loss | PEX14 CASZ1 |
nsv525693 | Chr.1:10634068 - 10780848 on Build GRCh38 | Loss | CASZ1 |
nsv828030 | Chr.1:10617505 - 10735104 on Build GRCh38 | Gain | PEX14 CASZ1 |
nsv953454 | Chr.1:10616244 - 10724643 on Build GRCh38 | Deletion | PEX14 CASZ1 |
nsv545401 | Chr.1:10630891 - 10697142 on Build GRCh38 | Loss | CASZ1 |
Set Membership: |
Intragenic Exonic DGV Variation |