Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23500 |
Gene Name: | dishevelled associated activator of morphogenesis 2 |
Gene Aliases: |
dJ90A20A.1 |
Location: |
Chr.6:39792366-39904877 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAAM2 | NM_001201427.1 | 25 | 4813 | NP_001188356.1 |
NM_015345.3 | 25 | 4874 | NP_056160.2 | |
XM_006715039.3 | 26 | 5485 | XP_006715102.3 | |
XM_006715040.3 | 25 | 5457 | XP_006715103.3 | |
XM_006715042.2 | 26 | 7480 | XP_006715105.1 | |
XM_006715043.2 | 26 | 4857 | XP_006715106.1 | |
XM_006715045.3 | 26 | 6072 | XP_006715108.1 | |
XM_006715046.3 | 26 | 5250 | XP_006715109.1 | |
XM_017010630.1 | 26 | 6096 | XP_016866119.1 | |
AB002379.2 | 25 | 4813 | ||
AL833083.2 | 25 | 4854 | CAD89973.1 | |
BX647839.1 | 9 | 5396 |
Set Membership: |
Intragenic Exonic non-DGV Variation |