Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | FOXP3 CACNA1F GAGE10 PPP1R3F CCDC22 |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | HDAC6 CACNA1F TFE3 WDR13 PQBP1 PCSK1N PRICKLE3 TIMM17B SUV39H1 EBP FOXP3 GPKOW TBC1D25 GRIPAP1 ERAS SLC35A2 CCDC22 WDR45 GLOD5 WAS PIM2 MAGIX RBM3 PRAF2 PPP1R3F OTUD5 LOC101927635 KCND1 SYP CCDC120 SYP-AS1 PORCN SLC38A5 GATA1 FTSJ1 PLP2 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | HDAC6 SSX9 CACNA1F TFE3 WDR13 SSX3 PQBP1 PCSK1N PRICKLE3 TIMM17B SUV39H1 SSX6 EBP SSX1 FOXP3 SSX4B GPKOW ZNF182 TBC1D25 SPACA5 GRIPAP1 SSX4 SPACA5B ERAS SLC35A2 CCDC22 WDR45 GLOD5 WAS ZNF630-AS1 PIM2 MAGIX RBM3 PRAF2 SSX5 PPP1R3F OTUD5 LOC101927635 KCND1 SYP CCDC120 ZNF630 SYP-AS1 PORCN SLC38A5 GATA1 FTSJ1 PLP2 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | FOXP3 CACNA1F GAGE10 PPP1R3F CCDC22 |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | FOXP3 CACNA1F CCDC22 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |