Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3636 |
Gene Name: | inositol polyphosphate phosphatase like 1 |
Gene Aliases: |
OPSMD, SHIP2 |
Location: |
Chr.11:72223861-72239147 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INPPL1 | NM_001567.3 | NP_001558.3 | ||
XM_005273978.3 | XP_005274035.1 | |||
XM_005273979.4 | XP_005274036.1 | |||
XM_011544999.2 | XP_011543301.1 | |||
XM_011545000.2 | XP_011543302.1 | |||
XM_017017673.1 | XP_016873162.1 | |||
BC140853.1 | ||||
L36818.1 | AAA96658.1 | |||
Y14385.1 | CAA74743.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951038 | Chr.11:72226157 - 72253456 on Build GRCh38 | Deletion | PHOX2A LOC220077 INPPL1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |