Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2692 |
Gene Name: | growth hormone releasing hormone receptor |
Gene Aliases: |
GHRFR, GRFR, IGHD1B |
Location: |
Chr.7:30964021-30979531 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRHR | NM_000823.3 | 4 | 364 | NP_000814.2 |
XM_011515263.1 | 1 | 382 | XP_011513565.1 | |
AF282259.1 | 1 | 382 | AAG15368.1 | |
AF282260.1 | 1 | 382 | AAG15369.1 | |
AF282261.1 | 2 | 206 | AAG15370.1 | |
AF282262.1 | 1 | 382 | AAG15371.1 | |
AY557192.1 | 4 | 316 | AAS59864.1 | |
L01406.1 | 4 | 367 | AAA35890.1 | |
L09237.1 | 4 | 364 | AAA58619.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain | GARS ADCYAP1R1 FAM188B CRHR2 GHRHR AQP1 INMT-FAM188B INMT |
nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain | GARS ADCYAP1R1 FAM188B CRHR2 GHRHR AQP1 INMT-FAM188B INMT |
esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain | GARS ADCYAP1R1 FAM188B CRHR2 GHRHR AQP1 INMT-FAM188B INMT |
nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion | NOD1 ADCYAP1R1 FAM188B LOC401320 DPY19L1P1 ZNRF2 MIR550A1 GARS AVL9 AQP1 INMT-FAM188B CCDC129 LINC01176 CRHR2 GHRHR MIR550B1 NEUROD6 PDE1C ZNRF2P1 LSM5 PPP1R17 LOC100130673 INMT GGCT DKFZP586I1420 LOC101928268 |
Set Membership: |
Intragenic Exonic DGV Variation |